Learn More
(EZ)-1-(4-Hydroxyphenyl)-1,2-diphenylbut-1-ene, Mikromol™
Product Code. 30051535
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051535 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051535 Supplier Mikromol Supplier No. MM0053.100025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(EZ)-1-(4-Hydroxyphenyl)-1,2-diphenylbut-1-ene
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (EZ)-1-(4-Hydroxyphenyl)-1,2-diphenylbut-1-ene
CAS 68684-63-9
Analyte or Component Names (EZ)-1-(4-Hydroxyphenyl)-1,2-diphenylbut-1-ene
Shipping Condition Room Temperature
Molecular Formula C22 H20 O
Certifications/Compliance ISO 17025
Synonym (EZ)-1-(4-Hydroxyphenyl)-1,2-diphenylbut-1-ene
InChI Formula InChI=1S/C22H20O/c1-2-21(17-9-5-3-6-10-17)22(18-11-7-4-8-12-18)19-13-15-20(23)16-14-19/h3-16,23H,2H2,1H3
SMILES CCC(=C(c1ccccc1)c2ccc(O)cc2)c3ccccc3
IUPAC Name 4-(1,2-diphenylbut-1-enyl)phenol
Molecular Weight (g/mol) 300.39
Quantity 25 mg
Formula Weight 300.1514 g/mol