Learn More
Memmert™ Extension of DAkkS Factory Calibration for IN110
Product Code. 17567365 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17567365
Brand: Memmert™ IN110E39697
Please to purchase this item. Need a web account? Register with us today!
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Incubators.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) IN110 Incubator
Product Type Factory Calibration Extension