Learn More
Etosalamide, Mikromol™
Product Code. 30051041
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 30051041
Brand: Mikromol MM1889.00
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Etosalamide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Etosalamide
CAS 15302-15-5
Analyte or Component Names Etosalamide
Shipping Condition Room Temperature
Molecular Formula C11 H15 N O3
Certifications/Compliance ISO 17025
Synonym Ethosalamide, Benzamide, 2-(2-ethoxyethoxy)-, Benzamide, o-(2-ethoxyethoxy)-, Ethosalamide, o-(2-Ethoxyethoxy)benzamide
InChI Formula InChI=1S/C11H15NO3/c1-2-14-7-8-15-10-6-4-3-5-9(10)11(12)13/h3-6H,2,7-8H2,1H3,(H2,12,13)
SMILES CCOCCOc1ccccc1C(=O)N
IUPAC Name 2-(2-ethoxyethoxy)benzamide
Molecular Weight (g/mol) 209.24
Quantity 100 mg
Formula Weight 209.1052 g/mol