Learn More
Etosalamide, LoGiCal
Product Code. 30463028
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30463028
Brand: LoGiCal LGCFOR1889.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Etosalamide
CAS 15302-15-5
Analyte or Component Names Etosalamide
Shipping Condition Room Temperature
Molecular Formula C11 H15 N O3
Certifications/Compliance ISO 9001
Synonym Ethosalamide, Benzamide, 2-(2-ethoxyethoxy)-, Benzamide, o-(2-ethoxyethoxy)-, Ethosalamide, o-(2-Ethoxyethoxy)benzamide
InChI Formula InChI=1S/C11H15NO3/c1-2-14-7-8-15-10-6-4-3-5-9(10)11(12)13/h3-6H,2,7-8H2,1H3,(H2,12,13)
Solution Type Neat
SMILES CCOCCOc1ccccc1C(=O)N
IUPAC Name 2-(2-ethoxyethoxy)benzamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 209.24
Quantity 10 mg
Formula Weight 209.1052 g/mol