Learn More
Etofenamate EP Impurity G, Mikromol™
Description
Etofenamate EP Impurity G
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxyethyl 2-[(3-(Trifluoro-methyl)phenyl)amino]benzoate |
| CAS | 32508-98-8 |
| Analyte or Component Names | 2-Hydroxyethyl 2-[(3-(Trifluoro-methyl)phenyl)amino]benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H14 F3 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Hydroxyethyl 2-[[3-(Trifluoromethyl)phenyl]amino]benzoate, 2-Hydroxyethyl 2-[[3-(Trifluoro-methyl)phenyl]amino]benzoate, Etofenamate Imp. G (EP), Etofenamate Impurity G |
| InChI Formula | InChI=1S/C16H14F3NO3/c17-16(18,19)11-4-3-5-12(10-11)20-14-7-2-1-6-13(14)15(22)23-9-8-21/h1-7,10,20-21H,8-9H2 |
| SMILES | OCCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F |
| IUPAC Name | 2-hydroxyethyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Show More |