Learn More
Ethyl Ester of Cetirizine Dihydrochloride, Mikromol™
Product Code. 17964456
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964456 100 mg 100mg
17961586 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17964456 Supplier Mikromol Supplier No. MM0380.15
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl Ester of Cetirizine Dihydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl Ester of Cetirizine Dihydrochloride
CAS 2732277-01-7
Analyte or Component Names Ethyl Ester of Cetirizine Dihydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C23 H29 Cl N2 O3 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Ethyl Ester of Cetirizine Dihydrochloride, (RS)-2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester dihydrochloride, Cetirizine Related Compound A as Dihydrochloride
InChI Formula InChI=1S/C23H29ClN2O3.2ClH/c1-2-29-22(27)18-28-17-16-25-12-14-26(15-13-25)23(19-6-4-3-5-7-19)20-8-10-21(24)11-9-20;;/h3-11,23H,2,12-18H2,1H3;2*1H
SMILES Cl.Cl.CCOC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name ethyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate;dihydrochloride
Molecular Weight (g/mol) 489.86
Quantity 100 mg
Formula Weight 488.14 g/mol