Learn More
Ethyl Ester of Cetirizine Dihydrochloride, Mikromol™
Description
Ethyl Ester of Cetirizine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Ethyl Ester of Cetirizine Dihydrochloride |
| CAS | 2732277-01-7 |
| Analyte or Component Names | Ethyl Ester of Cetirizine Dihydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C23 H29 Cl N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl Ester of Cetirizine Dihydrochloride, (RS)-2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester dihydrochloride, Cetirizine Related Compound A as Dihydrochloride |
| InChI Formula | InChI=1S/C23H29ClN2O3.2ClH/c1-2-29-22(27)18-28-17-16-25-12-14-26(15-13-25)23(19-6-4-3-5-7-19)20-8-10-21(24)11-9-20;;/h3-11,23H,2,12-18H2,1H3;2*1H |
| SMILES | Cl.Cl.CCOC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | ethyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate;dihydrochloride |
| Show More |