Learn More
Ethyl (5-Benzoyl-1H-benzimidazol-2-yl)carbamate, Mikromol™
Description
Ethyl (5-Benzoyl-1H-benzimidazol-2-yl)carbamate
Specifications
Specifications
| Chemical Name or Material | Ethyl (5-Benzoyl-1H-benzimidazol-2-yl)carbamate |
| CAS | 31430-19-0 |
| Analyte or Component Names | Ethyl (5-Benzoyl-1H-benzimidazol-2-yl)carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H15 N3 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethyl (5-Benzoyl-1H-benzimidazol-2-yl)carbamate, Mebendazole Imp. E (EP), Mebendazole Related Compound E, Mebendazole Impurity E |
| InChI Formula | InChI=1S/C17H15N3O3/c1-2-23-17(22)20-16-18-13-9-8-12(10-14(13)19-16)15(21)11-6-4-3-5-7-11/h3-10H,2H2,1H3,(H2,18,19,20,22) |
| SMILES | CCOC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3 |
| IUPAC Name | ethyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate |
| Show More |