missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate, Thermo Scientific™

Product Code. 11375579
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375579 1 g 1g
11385579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375579 Supplier Thermo Scientific Alfa Aesar Supplier No. H50504.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338954-49-7
Molecular Formula C14H14ClNO2
Molecular Weight (g/mol) 263.72
MDL Number MFCD00173354
InChI Key VTIBBHSQWPMQAQ-UHFFFAOYSA-N
Synonym ethyl 4-chloro-5,7-dimethyl-3-quinolinecarboxylate, acmc-1bmtw, ethyl4-chloro-5,7-dimethylquinoline-3-carboxylate, 4-chloro-5,7-dimethylquinoline-3-carboxylic acid ethyl ester
PubChem CID 1479090
IUPAC Name ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=C(C)C=C(C)C2=C1Cl

Specifications

Quantity 1 g
Formula Weight 263.73
Chemical Name or Material Ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.