accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COONCH3CH3Cl

CAS RN 338954-49-7

IUPAC Name: ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-5,7-dimethylquinoline-3-carboxylate
Molecular Weight (g/mol): 263.72
Molecular Formula: C14H14ClNO2
InChi Key: VTIBBHSQWPMQAQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=C(C)C=C(C)C2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results