missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate, 95%, Thermo Scientific™

Product Code. p-7032397
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15448665

Brand: Thermo Scientific Alfa Aesar H33837.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78185-92-9
Molecular Formula C7H9F6NO2
Molecular Weight (g/mol) 253.14
MDL Number MFCD00040845
InChI Key PGOSQXOLEQZPBM-UHFFFAOYNA-N
Synonym ethyl 4,4,4,4',4',4'-hexafluoro-dl-valinate, ethyl 2-amino-4,4,4-trifluoro-3-trifluoromethyl butanoate, ethyl 2s-2-amino-4,4,4-trifluoro-3-trifluoromethyl butanoate, 4,4,4,4',4',4'-hexafluorovaline, ethyl ester, ethyl 2-amino-4,4,4-trifluoro-3-trifluoromethyl b, ethyl 2-amino-4,4,4-trifluoro-3-trifluoromethyl butyrate, 2-amino-4,4,4-trifluoro-3-trifluoromethyl-butyric acid ethyl ester
PubChem CID 52991940
IUPAC Name ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoate
SMILES CCOC(=O)C(N)C(C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 70°C to 72°C (10 mmHg)
Refractive Index 1.36
Quantity 250 mg
Formula Weight 253.14
Percent Purity 95%
Chemical Name or Material Ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.