accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 78185-92-9

H3COOFFFFFFH2N

CAS RN 78185-92-9

IUPAC Name: ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoate
Synonyms: ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoate
Molecular Weight (g/mol): 253.14
Molecular Formula: C7H9F6NO2
InChi Key: PGOSQXOLEQZPBM-UHFFFAOYNA-N
SMILES: CCOC(=O)C(N)C(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)