missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(6-Amino-2,3-dichlorobenzyl)glycine Dihydrochloride(Anagrelide Impurity A), TRC

Product Code. 30381432
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381432 100 mg
30377046 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381432 Supplier Toronto Research Chemicals Supplier No. TRCE898180100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882301-57-7
Molecular Formula C11H16Cl4N2O2
Molecular Weight (g/mol) 350.07
Synonym N-[(6-Amino-2,3-dichlorophenyl)methyl]glycine Ethyl Ester Dihydrochloride, Ethyl (2-Amino-5,6-dichlorobenzyl)aminoacetate Dihydrochloride, Ethyl N-(6-Amino-2,3-dichlorobenzyl)glycine Dihydrochloride, USP Anagrelide Related Impurity A Dihydrochloride
IUPAC Name ethyl (6-amino-2,3-dichlorobenzyl)glycinate dihydrogen chloride
SMILES NC1=CC=C(Cl)C(Cl)=C1CNCC(OCC)=O.Cl.Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H14Cl2N2O2.2ClH/c1-2-17-10(16)6-15-5-7-9(14)4-3-8(12)11(7)13;;/h3-4,15H,2,5-6,14H2,1H3;2*1H
Formula Weight 348
Chemical Name or Material Ethyl 2-(6-Amino-2,3-dichlorobenzyl)glycine Dihydrochloride (Anagrelide Impurity A)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.