CAS RN 882301-57-7
CAS RN 882301-57-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 2-(6-Amino-2,3-dichlorobenzyl)glycine Dihydrochloride(Anagrelide Impurity A), TRC
CAS: 882301-57-7 Molecular Formula: C11H16Cl4N2O2 Molecular Weight (g/mol): 350.07 Synonym: N-[(6-Amino-2,3-dichlorophenyl)methyl]glycine Ethyl Ester Dihydrochloride,Ethyl (2-Amino-5,6-dichlorobenzyl)aminoacetate Dihydrochloride,Ethyl N-(6-Amino-2,3-dichlorobenzyl)glycine Dihydrochloride,USP Anagrelide Related Impurity A Dihydrochloride IUPAC Name: ethyl (6-amino-2,3-dichlorobenzyl)glycinate dihydrogen chloride SMILES: NC1=CC=C(Cl)C(Cl)=C1CNCC(OCC)=O.Cl.Cl