Learn More
Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate (Ethyl Ester of Diclofenac), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate (Ethyl Ester of Diclofenac)
Specifications
Specifications
| Chemical Name or Material | Diclofenac Ethyl Ester |
| CAS | 15307-77-4 |
| Analyte or Component Names | Diclofenac Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 Cl2 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl [2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Aceclofenac Imp. C (EP), Ethyl Ester of Diclofenac, Aceclofenac Impurity C |
| InChI Formula | InChI=1S/C16H15Cl2NO2/c1-2-21-15(20)10-11-6-3-4-9-14(11)19-16-12(17)7-5-8-13(16)18/h3-9,19H,2,10H2,1H3 |
| SMILES | CCOC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl |
| IUPAC Name | ethyl 2-[2-(2,6-dichloroanilino)phenyl]acetate |
| Show More |