Learn More
Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate (Ethyl Ester of Diclofenac), LoGiCal
Product Code. 30459192
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30459192
Brand: LoGiCal LGCFOR0006.11
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diclofenac Ethyl Ester
CAS 15307-77-4
Analyte or Component Names Diclofenac Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H15 Cl2 N O2
Certifications/Compliance ISO 9001
Synonym Ethyl [2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Aceclofenac Imp. C (EP), Ethyl Ester of Diclofenac, Aceclofenac Impurity C
InChI Formula InChI=1S/C16H15Cl2NO2/c1-2-21-15(20)10-11-6-3-4-9-14(11)19-16-12(17)7-5-8-13(16)18/h3-9,19H,2,10H2,1H3
Solution Type Neat
SMILES CCOC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name ethyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 324.2
Quantity 10 mg
Formula Weight 323.048 g/mol