Learn More
Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate, Mikromol™
Description
Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate
Specifications
Specifications
| Chemical Name or Material | Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate |
| CAS | 28018-10-2 |
| Analyte or Component Names | Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H23 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Pethidine Hydrochloride Imp. J (EP), Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate, Pethidine Imp. J (EP), Pethidine Hydrochloride Impurity J, Pethidine Impurity J |
| InChI Formula | InChI=1S/C16H23NO2/c1-3-17-12-10-16(11-13-17,15(18)19-4-2)14-8-6-5-7-9-14/h5-9H,3-4,10-13H2,1-2H3 |
| SMILES | CCOC(=O)C1(CCN(CC)CC1)c2ccccc2 |
| IUPAC Name | ethyl 1-ethyl-4-phenylpiperidine-4-carboxylate |
| Show More |