Learn More
Ethephon-hydroxy 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30315614
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315614 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30315614 Supplier Dr. Ehrenstorfer Supplier No. DREA13230200AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethephon-hydroxy
CAS 22987-21-9
Analyte or Component Names Ethephon-hydroxy
Shipping Condition Room Temperature
Molecular Formula C2 H7 O4 P
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Ethephon-hydroxy
InChI Formula InChI=1S/C2H7O4P/c3-1-2-7(4,5)6/h3H,1-2H2,(H2,4,5,6)
SMILES OCCP(=O)(O)O
IUPAC Name 2-hydroxyethylphosphonic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 126.05
Quantity 1 mL
Formula Weight 126.0082 g/mol