Learn More
Erbium AAS Standard, 1000 mg/L Er in HNO3 2%, Honeywell Fluka™
Brand: Honeywell Fluka E4025-100ML
Additional Details : Weight : 0.10100kg ??product.attributeSet.Transport?? : UN number : 3264 Chem class : 8 Pack group : III
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Erbium AAS Standard | |
1.01 g/cm3 | |
100 mL |
Standards and Modifiers for AAS | |
<2 |