Learn More
Memmert™ Entry Port 57 mm for UN750plus
Product Code. 17520446 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
57 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17520446 57 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17520446 Supplier Memmert™ Supplier No. UN750plusF8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UN750plus Oven
Type Entry Port
Diameter (Metric) 57 mm