Learn More
Memmert™ Entry Port 57 mm for UF450plus
Product Code. 17509405 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
57 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17509405 57 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17509405 Supplier Memmert™ Supplier No. UF450plusF8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UF450plus Oven
Type Entry Port
Diameter (Metric) 57 mm