Learn More
Memmert™ Entry Port 57 mm for UF110plus
Product Code. 17558975 Shop All Memmert Products
Click to view available options
Diameter (Metric):
57 mm
Unit Size:
Each
Product Code. 17558975
Brand: Memmert™ UF110plusF8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UF110plus Oven
Type Entry Port
Diameter (Metric) 57 mm