Learn More
Memmert™ Entry Port 38 mm for UN260plus
Shop All Memmert Products
Click to view available options
Diameter (Metric):
38 mm
Unit Size:
Each
Description
- Entry port with 38 mm clear diameter
- Flap and stopper closure
- Custom position TBC in the back
Specifications
Specifications
| For Use With (Equipment) | UN260plus Oven |
| Type | Entry Port |
| Diameter (Metric) | 38 mm |