Learn More
Memmert™ Entry Port 23 mm for IN110, Custom TBC Back Side
Product Code. 17537375 Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17537375
Brand: Memmert™ IN110F6
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Optional entry port for Memmert Incubators.
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IN110 Incubator
Type Entry Port
Diameter (Metric) 23 mm