Learn More
Memmert™ Entry Port 100 mm for UN110plus
Product Code. 17549825 Shop All Memmert Products
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Each
Product Code. 17549825
Brand: Memmert™ UN110plusF9
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 100 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UN110plus Oven
Type Entry Port
Diameter (Metric) 100 mm