Learn More
Enrofloxacin N-Oxide Hydrochloride, Mikromol™
Description
Enrofloxacin N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Enrofloxacin N-Oxide Hydrochloride |
| CAS | 2731368-88-8 |
| Analyte or Component Names | Enrofloxacin N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 F N3 O4 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Enrofloxacin N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C19H22FN3O4.ClH/c1-2-23(27)7-5-21(6-8-23)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-22(16)12-3-4-12;/h9-12H,2-8H2,1H3,(H,25,26);1H |
| SMILES | Cl.CC[N+]1([O-])CCN(CC1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4 |
| IUPAC Name | 1-cyclopropyl-7-(4-ethyl-4-oxidopiperazin-4-ium-1-yl)-6-fluoro-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| Show More |