Learn More
Enrofloxacin Isopropyl Ester, Mikromol™
Description
Enrofloxacin Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Enrofloxacin Isopropyl Ester |
| CAS | 2724726-50-3 |
| Analyte or Component Names | Enrofloxacin Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 F N3 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Enrofloxacin Isopropyl Ester |
| InChI Formula | InChI=1S/C22H28FN3O3/c1-4-24-7-9-25(10-8-24)20-12-19-16(11-18(20)23)21(27)17(22(28)29-14(2)3)13-26(19)15-5-6-15/h11-15H,4-10H2,1-3H3 |
| SMILES | CCN1CCN(CC1)c2cc3N(C=C(C(=O)OC(C)C)C(=O)c3cc2F)C4CC4 |
| IUPAC Name | propan-2-yl 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxoquinoline-3-carboxylate |
| Show More |