missing translation for 'onlineSavingsMsg'
Learn More

EG00229, MedChemExpress

Product Code. 16495126
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495126

Brand: Medchem Express HY10799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EG00229 is a neuropilin 1 (NRP1) receptor antagonist. EG00229 selectively inhibits VEGF-A binding to NRP1 b1 domain with an IC50 of 3 μM, but has no effect on VEGFA binding to VEGFR-1 and VEGFR-2.

EG00229 is a neuropilin 1 (NRP1) receptor antagonist. EG00229 selectively inhibits VEGF-A binding to NRP1 b1 domain with an IC50 of 3 μM, but has no effect on VEGFA binding to VEGFR-1 and VEGFR-2.

Specifications

Chemical Name or Material EG00229
CAS 1210945-69-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H20F3N7O7S3
Quantity 100 mg
Solubility Information DMSO : ≥ 41.4 mg/mL (67.69 mM)
SMILES N=C(N)NCCC[C@@H](C(O)=O)NC(C1=C(NS(=O)(C2=CC=CC3=NSN=C32)=O)C=CS1)=O.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 611.6
Formula Weight 611.6
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.