missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21148-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcriptional activator which recognizes variations of the palindromic sequence 5′-ATTCCCNNGGGAATT-3′.Specifications
| EBF3 | |
| Polyclonal | |
| Unconjugated | |
| EBF3 | |
| COE3, early B cell factor 3, EBF 3, EBF3, O/E 2, OE 2, Olf 1/EBF like 2, Transcription factor COE3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13593, 253738, 361668 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08791, Q9H4W6 | |
| Ebf3 | |
| EBF3 Fusion Protein Ag15693 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction