missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | EBF3 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870755
|
Proteintech
21148-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860755
|
Proteintech
21148-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transcriptional activator which recognizes variations of the palindromic sequence 5′-ATTCCCNNGGGAATT-3′.Specifications
| EBF3 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O08791, Q9H4W6 | |
| 13593, 253738, 361668 | |
| EBF3 Fusion Protein Ag15693 | |
| Primary | |
| -20°C | |
| EBF3 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| COE3, early B cell factor 3, EBF 3, EBF3, O/E 2, OE 2, Olf 1/EBF like 2, Transcription factor COE3 | |
| Ebf3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title