Learn More
Dubermatinib, MedChemExpress
Product Code. 16481627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16481627
Brand: Medchem Express HY12963
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Dubermatinib (TP-0903) is a potent and selective Axl receptor tyrosine kinase inhibitor with an IC50 value of 27 nM.
Dubermatinib (TP-0903) is a potent and selective Axl receptor tyrosine kinase inhibitor with an IC50 value of 27 nM.
Specifications
Chemical Name or Material Dubermatinib
CAS 1341200-45-0
Color White
Purity Grade Notes Research
Molecular Formula C24H30ClN7O2S
Quantity 50 mg
Synonym TP-0903
Solubility Information DMSO : 3.33 mg/mL (6.45 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1NC2=NC(NC3=CC=C(CN4CCN(C)CC4)C=C3)=NC=C2Cl)(N(C)C)=O
Molecular Weight (g/mol) 516.06
Formula Weight 516.06
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease