Learn More
Dräger™ Short-Term Detector Tubes - Phosgene
Product Code. 11368291
Click to view available options
Components Measured:
Phosgene 0.02/a
Phosgene 0.25/b
Measurement Range:
0.02 to 1ppm
0.25 to 5ppm
Unit Size:
Each
Pack of 10
Product Code. 11368291
Brand: Dräger™ 8101521
Please to purchase this item. Need a web account? Register with us today!
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Phosgene 0.02/a
Humidity Range 3 to 15mg/L
Measurement Range 0.02 to 1ppm
No. of Strokes 20/40
Quantity 10
Response Time 6 to 12 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
compliance-icons
Product Identifier
  • DR PHOSGEN 0,02/A - 8101521
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contains : phosgene