Learn More
Doxylamine N-Oxide Dihydrochloride, Mikromol™
Description
Doxylamine N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Doxylamine N-Oxide Dihydrochloride |
| CAS | 2724689-64-7 |
| Analyte or Component Names | Doxylamine N-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H22 N2 O2 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Doxylamine N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C17H22N2O2.2ClH/c1-17(15-9-5-4-6-10-15,16-11-7-8-12-18-16)21-14-13-19(2,3)20;;/h4-12H,13-14H2,1-3H3;2*1H |
| SMILES | Cl.Cl.CC(OCC[N+](C)(C)[O-])(c1ccccc1)c2ccccn2 |
| IUPAC Name | N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine oxide;dihydrochloride |
| Show More |