Learn More
Doxepin N-Oxide Hydrochloride, Mikromol™
Product Code. 30052134
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052134 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052134 Supplier Mikromol Supplier No. MM0032.120025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Doxepin N-Oxide Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Doxepin N-Oxide Hydrochloride
CAS 2734342-87-9
Analyte or Component Names Doxepin N-Oxide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym 9,11-Didehydroestriol, Doxepin N-Oxide Hydrochloride
InChI Formula InChI=1S/C11H9Cl4NO2/c12-5-8(16-11(18)10(14)15)9(17)6-1-3-7(13)4-2-6/h1-4,8,10H,5H2,(H,16,18)
SMILES Cl.C[N+](C)([O-])CC\C=C\1/c2ccccc2COc3ccccc13
IUPAC Name (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine oxide;hydrochloride
Molecular Weight (g/mol) 331.84
Quantity 25 mg
Formula Weight 331.1339 g/mol