Learn More
Doxapram N-Oxide, Mikromol™
Description
Doxapram N-Oxide
Specifications
Specifications
| Chemical Name or Material | Doxapram N-Oxide |
| CAS | 2724726-82-1 |
| Analyte or Component Names | Doxapram N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H30 N2 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Doxapram N-Oxide |
| InChI Formula | InChI=1S/C24H30N2O3/c1-2-25-19-22(13-14-26(28)15-17-29-18-16-26)24(23(25)27,20-9-5-3-6-10-20)21-11-7-4-8-12-21/h3-12,22H,2,13-19H2,1H3 |
| SMILES | CCN1CC(CC[N+]2([O-])CCOCC2)C(C1=O)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 1-ethyl-4-[2-(4-oxidomorpholin-4-ium-4-yl)ethyl]-3,3-diphenylpyrrolidin-2-one |
| Show More |