Learn More
Dobutamine EP Impurity C (as Hydrochloride), Mikromol™
Description
Dobutamine EP Impurity C (as Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | (2RS)-N-[2-(3,4-Dimethoxyphenyl)ethyl]-4-(4-methoxyphenyl)butan-2-amine Hydrochloride |
| CAS | 51062-14-7 |
| Analyte or Component Names | (2RS)-N-[2-(3,4-Dimethoxyphenyl)ethyl]-4-(4-methoxyphenyl)butan-2-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H29 N O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (3'R)-Ezetimibe, Dobutamine Hydrochloride Imp. C (EP), Dobutamine Imp. C (EP), N-[2-(3,4-Dimethoxyphenyl)ethyl]-4-methoxy-alpha-methylbenzenepropanamine Hydrochloride (1:1), (2RS)-N-[2-(3,4-Dimethoxyphenyl)-ethyl]-4-(4-methoxyphenyl)-butan-2-amine Hydrochloride, Benzenepropanamine, N-[2-(3,4-dimethoxyphenyl)ethyl]-4-methoxy-alpha-methyl-, hydrochloride (9CI), Trimethoxy Dobutamine Hydrochloride, Dobutamine Related Compound C as Hydrochloride, Dobutamine Hydrochloride Impurity C as Hydrochloride, Dobutamine Impurity C as Hydrochloride |
| Solution Type | Neat |
| SMILES | Cl.COc1ccc(CCC(C)NCCc2ccc(OC)c(OC)c2)cc1 |
| Molecular Weight (g/mol) | 379.93 |
| Show More |