Learn More
DNPH Mixture 749 20 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 958-60-1, 1019-57-4, 1567-89-1, 888-54-0, 1157-84-2, 1527-98-6 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |