missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
218.00 €
Specifications
| Antigen | DNMT3B |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2.0 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DNMT3B Polyclonal antibody specifically detects DNMT3B in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Knockdown.Specifications
| DNMT3B | |
| Western Blot 2.0 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB | |
| DNMT3B | |
| IgG | |
| Affinity Purified | |
| This detects DNA methyltransferase 3b (Dnmt3b) from human and mouse tissues and cells as well as recombinant human Dnmt3b. This detects, to a lesser extent, full-length human recombinant Dnmt3a. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| O88509 | |
| 1789 | |
| Synthetic peptide corresponding to the residues M(1) K G D S R H L N E E E G A(14) C of mouse DNMT3b | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title