missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-516-0.025mg
This item is not returnable.
View return policy
Description
DNMT3B Polyclonal antibody specifically detects DNMT3B in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Knockdown.
Specifications
| DNMT3B | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This detects DNA methyltransferase 3b (Dnmt3b) from human and mouse tissues and cells as well as recombinant human Dnmt3b. This detects, to a lesser extent, full-length human recombinant Dnmt3a. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2.0 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated | |
| O88509 | |
| DNMT3B | |
| Synthetic peptide corresponding to the residues M(1) K G D S R H L N E E E G A(14) C of mouse DNMT3b | |
| 0.025 mg | |
| Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics | |
| 1789 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction