missing translation for 'onlineSavingsMsg'
Learn More

DNAJC15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen DNAJC15
Concentration 0.21 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16815544
View Documents
Proteintech
16063-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16805544
View Documents
Proteintech
16063-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNAJC15 is a negative regulator of the mitochondrial respiratory chain. It prevents mitochondrial hyperpolarization state and restricts mitochondrial generation of ATP. It acts as an import component of the TIM23 translocase complex. It stimulates the ATPase activity of HSPA9.
Specifications

Specifications

DNAJC15
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Rat, Mouse
Q78YY6, Q9Y5T4
290370, 29103, 66148
DNAJC15 Fusion Protein Ag8911
Primary
-20°C
DNAJC15
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DNAJC15, DNAJD1, HSD18, MCJ
Dnajc15
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.