missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16063-1-AP-150UL
This item is not returnable.
View return policy
Description
DNAJC15 is a negative regulator of the mitochondrial respiratory chain. It prevents mitochondrial hyperpolarization state and restricts mitochondrial generation of ATP. It acts as an import component of the TIM23 translocase complex. It stimulates the ATPase activity of HSPA9.Specifications
| DNAJC15 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC15 | |
| DNAJC15, DNAJD1, HSD18, MCJ | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 290370, 29103, 66148 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q78YY6, Q9Y5T4 | |
| Dnajc15 | |
| DNAJC15 Fusion Protein Ag8911 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction