Learn More
Disopyramide Impurity A (as Dihydrogen Phosphate), Mikromol™
Description
Disopyramide Impurity A (as Dihydrogen Phosphate)
Specifications
Specifications
| Chemical Name or Material | Diisopyronitrile Dihydrogen Phosphate |
| CAS | 2724689-67-0 |
| Analyte or Component Names | Diisopyronitrile Dihydrogen Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N3 . H3 O4 P |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-4-[Bis(1-methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanenitrile Dihydrogen Phosphate (Diisopyronitrile Dihydrogen Phosphate), Disopyramide Imp. A (EP), (2RS)-4-[bis(1-methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanenitrile dihydrogen phosphate (di-isopyronitrile dihydrogen phosphate), Disopyramide Impurity A as Dihydrogen Phosphate |
| InChI Formula | InChI=1S/C21H27N3.H3O4P/c1-17(2)24(18(3)4)15-13-21(16-22,19-10-6-5-7-11-19)20-12-8-9-14-23-20;1-5(2,3)4/h5-12,14,17-18H,13,15H2,1-4H3;(H3,1,2,3,4) |
| SMILES | CC(C)N(CCC(C#N)(c1ccccc1)c2ccccn2)C(C)C.OP(=O)(O)O |
| IUPAC Name | 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanenitrile;phosphoric acid |
| Show More |