Learn More
Disodium [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]-N-bis(methanesulfonate), Mikromol
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Disodium [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]-N-bis(methanesulfonate), Mikromol
Specifications
Specifications
| Chemical Name or Material | Disodium [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]-N-bis(methanesulfonate) |
| Analyte or Component Names | Disodium [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]-N-bis(methanesulfonate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 N3 O7 S2 . 2 Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Disodium [(1,5-Dimethyl-3-oxo-2-phenyl-pyrazol-4-yl)-(sulfonatomethyl)amino]methanesulfonate |
| InChI Formula | InChI=1S/C13H17N3O7S2.2Na/c1-10-12(15(8-24(18,19)20)9-25(21,22)23)13(17)16(14(10)2)11-6-4-3-5-7-11;;/h3-7H,8-9H2,1-2H3,(H,18,19,20)(H,21,22,23);;/q;2*+1/p-2 |
| Solution Type | Neat |
| SMILES | [Na+].[Na+].CN1N(C(=O)C(=C1C)N(CS(=O)(=O)[O-])CS(=O)(=O)[O-])c2ccccc2 |
| IUPAC Name | disodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-(sulfonatomethyl)amino]methanesulfonate |
| Show More |