missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethanol (Benzhydrol), Mikromol™

Product Code. p-200130016
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17971316

Brand: Mikromol MM0490.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diphenylmethanol (Benzhydrol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylmethanol
CAS 91-01-0
Analyte or Component Names Diphenylmethanol
Shipping Condition Room Temperature
Molecular Formula C13 H12 O
Certifications/Compliance ISO 17025
Synonym Cyclizine Hydrochloride Imp. B (EP), Cyclizine Imp. B (EP), Dimenhydrinate Imp. I (EP), Diphenhydramine Hydrochloride Imp. D (EP), Diphenhydramine Imp. D (EP), Ebastine Imp. A (EP), Diphenylmethanol, Benzhydrol, Cyclizine Hydrochloride Impurity B, Dimenhydrinate Impurity I, Diphenhydramine Hydrochloride Impurity D, Ebastine Impurity A, Cyclizine Impurity B, Diphenhydramine Impurity D
InChI Formula InChI=1S/C13H12O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13-14H
SMILES OC(c1ccccc1)c2ccccc2
IUPAC Name diphenylmethanol
Molecular Weight (g/mol) 184.23
Quantity 25 mg
Formula Weight 184.0888 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.