Learn More
Diphenylcarbamic Chloride, Mikromol™
Product Code. 30052201
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052201 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052201 Supplier Mikromol Supplier No. MM1480.010025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diphenylcarbamic Chloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diphenylcarbamic Chloride
CAS 83-01-2
Analyte or Component Names Diphenylcarbamic Chloride
Shipping Condition Room Temperature
Molecular Formula C13 H10 Cl N O
Certifications/Compliance ISO 17025
Synonym Diphenylcarbamic Chloride, Temozolomide Related Compound C (USP), Temozolomide Related Compound C
InChI Formula InChI=1S/C13H10ClNO/c14-13(16)15(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
SMILES ClC(=O)N(c1ccccc1)c2ccccc2
IUPAC Name N,N-diphenylcarbamoyl chloride
Molecular Weight (g/mol) 231.68
Quantity 25 mg
Formula Weight 231.0451 g/mol