Learn More
Diphenidol Hydrochloride, TRC
Product Code. 30136466
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136466 1 g 1g
30137119 50 mg 50mg
30010562 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136466 Supplier Toronto Research Chemicals Supplier No. TRCD4869401G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3254-89-5
Molecular Formula C21 H27 N O . Cl H
Molecular Weight (g/mol) 345.91
Synonym 1-Piperidinebutanol, α,α-diphenyl-, hydrochloride (1:1), 1-Piperidinebutanol, α,α-diphenyl-, hydrochloride (6CI,7CI,8CI,9CI), Ansmin, Cefadol, Celmidol, Difenidol hydrochloride, Difenidolin, Diphenidol hydrochloride, Maniol, Mecalmin, Pineroro, SKF 478A, Satanolon, Tenesdol, Vontrol, Wansar
IUPAC Name 1,1-diphenyl-4-piperidin-1-ylbutan-1-ol;hydrochloride
SMILES Cl.OC(CCCN1CCCCC1)(c2ccccc2)c3ccccc3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C21H27NO.ClH/c23-21(19-11-4-1-5-12-19,20-13-6-2-7-14-20)15-10-18-22-16-8-3-9-17-22;/h1-2,4-7,11-14,23H,3,8-10,15-18H2;1H
Formula Weight 345.1859
Chemical Name or Material Difenidol Hydrochloride