missing translation for 'onlineSavingsMsg'
Learn More

Dimethylmethoxy(3,3,3-trifluoropropyl)silane, 98%, Thermo Scientific Chemicals

Product Code. 11348586
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348586 1 g 1g
11358586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348586 Supplier Thermo Scientific Alfa Aesar Supplier No. L17642.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4852-13-5
Molecular Formula C6H13F3OSi
Molecular Weight (g/mol) 186.25
MDL Number MFCD00236726
InChI Key QRLCLBZGWYJTHP-UHFFFAOYSA-N
Synonym dimethylmethoxy 3,3,3-trifluoropropyl silane, methoxydimethyl 3,3,3-trifluoropropyl silane, acmc-20al5w, 3,3,3-trifluoropropyl dimethyl methoxysilane, methoxy dimethyl 3,3,3-trifluoropropyl silane
PubChem CID 22497830
IUPAC Name methoxydimethyl(3,3,3-trifluoropropyl)silane
SMILES CO[Si](C)(C)CCC(F)(F)F

Specifications

Quantity 1 g
UN Number UN1993
Formula Weight 186.25
Percent Purity 98%
Refractive Index 1.359
Sensitivity Moisture sensitive
Chemical Name or Material Dimethylmethoxy(3,3,3-trifluoropropyl)silane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.