CAS RN 4852-13-5
CAS RN 4852-13-5
IUPAC Name:
methoxydimethyl(3,3,3-trifluoropropyl)silane
Synonyms:
methoxydimethyl(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol):
186.25
Molecular Formula:
C6H13F3OSi
InChi Key:
QRLCLBZGWYJTHP-UHFFFAOYSA-N
SMILES:
CO[Si](C)(C)CCC(F)(F)F
1
–
1
of
1
results
1
Dimethylmethoxy(3,3,3-trifluoropropyl)silane, 98%, Thermo Scientific Chemicals
CAS: 4852-13-5 Molecular Formula: C6H13F3OSi Molecular Weight (g/mol): 186.25 MDL Number: MFCD00236726 InChI Key: QRLCLBZGWYJTHP-UHFFFAOYSA-N Synonym: dimethylmethoxy 3,3,3-trifluoropropyl silane,methoxydimethyl 3,3,3-trifluoropropyl silane,acmc-20al5w,3,3,3-trifluoropropyl dimethyl methoxysilane,methoxy dimethyl 3,3,3-trifluoropropyl silane PubChem CID: 22497830 IUPAC Name: methoxydimethyl(3,3,3-trifluoropropyl)silane SMILES: CO[Si](C)(C)CCC(F)(F)F