Learn More
Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Product Code. 30462938
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462938 4 x 25 mg 100mg
30049936 100 mg 100mg
30052518 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30462938 Supplier Mikromol Supplier No. MM0139.0200254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 91189-59-2
Analyte or Component Names Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Shipping Condition Room Temperature
Molecular Formula C17 H17 Cl2 N O4
Certifications/Compliance ISO 17025
Synonym Felodipine Imp. B (EP), Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Felodipine Impurity B
InChI Formula InChI=1S/C17H17Cl2NO4/c1-8-12(16(21)23-3)14(10-6-5-7-11(18)15(10)19)13(9(2)20-8)17(22)24-4/h5-7,14,20H,1-4H3
Solution Type Neat
SMILES COC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)OC)C
IUPAC Name dimethyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 370.23
Quantity 4 x 25 mg
Formula Weight 369.0535 g/mol