Learn More
Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Description
Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Specifications
Specifications
| Chemical Name or Material | Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| CAS | 91189-59-2 |
| Analyte or Component Names | Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 Cl2 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Felodipine Imp. B (EP), Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Felodipine Impurity B |
| InChI Formula | InChI=1S/C17H17Cl2NO4/c1-8-12(16(21)23-3)14(10-6-5-7-11(18)15(10)19)13(9(2)20-8)17(22)24-4/h5-7,14,20H,1-4H3 |
| SMILES | COC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)OC)C |
| IUPAC Name | dimethyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Show More |